Merck
  • Further calculations on solubility of 3-amino-1-adamantanol in ethanol+ water binary solvent mixtures at various temperatures.
Further calculations on solubility of 3-amino-1-adamantanol in ethanol+ water binary solvent mixtures at various temperatures.
Journal of Molecular Liquids (2016)
Jouyban A, et al.