Merck
17β-estradiol (E2) in membranes: Orientation and dynamic properties.
Biochimica et biophysica acta (2015-11-27)
Jason J Guo, De-Ping Yang, Xiaoyu Tian, V Kiran Vemuri, Dali Yin, Chen Li, Richard I Duclos, Lingling Shen, Xiaoyu Ma, David R Janero, Alexandros Makriyannis
ZUSAMMENFASSUNG
Non-genomic membrane effects of estrogens are of great interest because of the diverse biological activities they may elicit. To further our understanding of the molecular features of the interaction between estrogenic hormones and membrane bilayers, we have determined the preferred orientation, location, and dynamic properties of 17β-estradiol (E2) in two different phospholipid membrane environments using (2)H-NMR and 2D (1)H-(13)C HSQC in conjunction with molecular dynamics simulations. Unequivocal spectral assignments to specific (2)H labels were made possible by synthesizing six selectively deuterated E2 molecules. The data allow us to conclude that the E2 molecule adopts a nearly "horizontal" orientation in the membrane bilayer with its long axis essentially perpendicular to the lipid acyl-chains. All four rings of the E2 molecule are located near the membrane interface, allowing both the E2 3-OH and the 17β-OH groups to engage in hydrogen bonding and electrostatic interactions with polar phospholipid groups. The findings augment our knowledge of the molecular interactions between E2 and membrane bilayer and highlight the asymmetric nature of the dynamic motions of the rigid E2 molecule in a membrane environment.