Merck
  • Microparticulate β-glucan vaccine conjugates phagocytized by dendritic cells activate both naïve CD4 and CD8 T cells in vitro.
Microparticulate β-glucan vaccine conjugates phagocytized by dendritic cells activate both naïve CD4 and CD8 T cells in vitro.
Cellular immunology (2015-11-10)
Vanessa K Berner, Sally A duPre, Doug Redelman, Kenneth W Hunter
ZUSAMMENFASSUNG
Microparticulate β-glucan (MG) conjugated to vaccine antigen has been shown to serve as an effective adjuvant in vivo. To further study antigen presentation by MG:vaccine conjugates, bone marrow-derived dendritic cells (BMDC) were treated with MG conjugated to ovalbumin (OVA), then interacted with splenocytes from DO11.10 transgenic mice expressing an OVA peptide-specific T cell receptor. BMDC treated with MG:OVA induced significantly higher numbers of activated (CD25+CD69+) OVA-specific CD4+ T cells than BMDC treated with OVA alone. BMDC treated with MG:OVA upregulated CD86 and CD40 expression as well as MG alone, indicating that conjugation of OVA does not alter the immunostimulatory capacity of MG. Activation of CD8+ OVA-specific OT-1 cells showed that MG:OVA is also capable of enhancing cross-presentation by BMDC to CD8+ cytotoxic T cells. These results show that MG acts as an adjuvant to enhance antigen presentation by dendritic cells to naïve, antigen-specific CD4 and CD8 T cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethyl-carbodiimid, ≥97.0% (T)