Merck
  • Comparison of alkali treatments for efficient release of p-coumaric acid and enzymatic saccharification of sorghum pith.
Comparison of alkali treatments for efficient release of p-coumaric acid and enzymatic saccharification of sorghum pith.
Bioresource technology (2016-02-13)
Kankan Jiang, Lulu Li, Liangkun Long, Shaojun Ding
ZUSAMMENFASSUNG
Two separate temperature and time ranges were respectively conducted for optimizing release of p-coumaric acid and enzymatic saccharification of sorghum pith by NaOH pretreatment using response surface methodology. Two desirable pretreatment conditions were selected as follows: 37°C, 2% NaOH and 12h, and 100°C, 1.75% NaOH and 37min in the low and high temperature ranges, respectively. Under these conditions, the enzymatic glucose yields were 85.6% and 90.4% respectively, whereas p-coumaric acid yields were 95.1% and 98.1% respectively. The final recovery of esterified p-coumaric acid reached 82.8% and 87.4% respectively after further separation with HP-20 resin. Interestingly, strong linear correlations exist between p-coumaric acid release with glucan saccharification yield and lignin dissolution. These results indicate that sorghum pith could be an attractive source for natural p-coumaric acid and efficient release of p-coumaric acid and enzymatic saccharification of sorghum pith can be achieved by mild NaOH pretreatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cellulase aus Trichoderma reesei, aqueous solution, ≥700 units/g