Merck
  • APPL proteins promote TGFβ-induced nuclear transport of the TGFβ type I receptor intracellular domain.
APPL proteins promote TGFβ-induced nuclear transport of the TGFβ type I receptor intracellular domain.
Oncotarget (2015-11-20)
Jie Song, Yabing Mu, Chunyan Li, Anders Bergh, Marta Miaczynska, Carl-Henrik Heldin, Marene Landström
ZUSAMMENFASSUNG
The multifunctional cytokine transforming growth factor-β (TGFβ) is produced by several types of cancers, including prostate cancer, and promote tumour progression in autocrine and paracrine manners. In response to ligand binding, the TGFβ type I receptor (TβRI) activates Smad and non-Smad signalling pathways. The ubiquitin-ligase tumour necrosis factor receptor-associated factor 6 (TRAF6) was recently linked to regulate intramembrane proteolytic cleavage of the TβRI in cancer cells. Subsequently, the intracellular domain (ICD) of TβRI enters in an unknown manner into the nucleus, where it promotes the transcription of pro-invasive genes, such as MMP2 and MMP9. Here we show that the endocytic adaptor molecules APPL1 and APPL2 are required for TGFβ-induced nuclear translocation of TβRI-ICD and for cancer cell invasiveness of human prostate and breast cancer cell lines. Moreover, APPL proteins were found to be expressed at high levels in aggressive prostate cancer tissues, and to be associated with TβRI in a TRAF6-dependent manner. Our results suggest that the APPL-TβRI complex promotes prostate tumour progression, and may serve as a prognostic marker.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SB−3CT, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Appl1