Merck
  • Modification of rubber wood with styrene in combination with diethyl allyl phosphate as the flame-retardant agent.
Modification of rubber wood with styrene in combination with diethyl allyl phosphate as the flame-retardant agent.
Journal of Applied Polymer Science (2007)
Devi RR, et al.