Merck
  • Heterocyclic analogs of nucleosides: synthesis and biological evaluation of novel analogs of puromycin.
Heterocyclic analogs of nucleosides: synthesis and biological evaluation of novel analogs of puromycin.
Canadian Journal of Chemistry (1994)
Hultin P G and Walter A S