Merck
  • Selective oxidation of optically active sec,sec-1,2-diols by dioxiranes. A practical method for the synthesis of homochiral .alpha.-hydroxy ketones in high optical purity
Selective oxidation of optically active sec,sec-1,2-diols by dioxiranes. A practical method for the synthesis of homochiral .alpha.-hydroxy ketones in high optical purity
The Journal of Organic Chemistry (1993)
Lucia D'Accolti, et al.