Merck
  • The human hedgehog-interacting protein gene: structure and chromosome mapping to 4q31.21-->q31.3.
The human hedgehog-interacting protein gene: structure and chromosome mapping to 4q31.21-->q31.3.
Cytogenetics and cell genetics (2001-07-04)
M Bak, C Hansen, K Friis Henriksen, N Tommerup
ZUSAMMENFASSUNG
Hedgehog-interacting protein (Hhip) is a novel regulatory component in the vertebrate hedgehog-signalling pathway. The murine Hhip encodes a type I TM protein that attenuates hedgehog signalling by binding all three mammalian hedgehog proteins. Here we describe the cloning and characterisation of the homologous human hedgehog-interacting protein gene (HHIP). HHIP comprises 13 exons and spans >91kb encoding a protein of 700 aa which shares 94% sequence iden- tity with mouse Hhip. HHIP maps to chromosome 4q31.21--> q31.3. Additionally, we have mapped murine Hhip to chromosome 8.