Merck
Exportin 4 interacts with Sox9 through the HMG Box and inhibits the DNA binding of Sox9.
PloS one (2011-10-13)
Megumi Tsuchiya, Hidesato Ogawa, Taiga Suzuki, Noriyuki Sugiyama, Tokuko Haraguchi, Yasushi Hiraoka
ZUSAMMENFASSUNG
Sox9 is a transcription factor that is required for tissue development in mammals. In general, such transcription factors require co-regulators for precise temporal and spatial control of the activation and inactivation of the numerous genes necessary for precise development during embryogenesis. Here we identify a new Sox9 co-regulator: Using affinity chromatography with immobilized Sox9 protein, we identified exportin 4 (Exp4) as an interacting protein of Sox9 in human cultured cells. Interaction between endogenous Exp4 and Sox9 proteins was confirmed in the human osteosarcoma U2OS cells by immunoprecipitation experiments using anti-Sox9 antibody. siRNA depletion of Exp4 enhanced transcription of Sox9 target genes in U2OS cells, but did not affect nuclear localization of Sox9. These results suggest that Exp4 regulates Sox9 activity in the nucleus. Furthermore we found that the HMG box of Sox9 was responsible for binding to Exp4, and the HMG box was required for suppression of Sox9-mediated transcription. This contrasts with the known Sox9 co-regulators which bind to its transcriptional activation domain. Chromatin immunoprecipitation analyses revealed that Exp4 prevents Sox9 binding to the enhancers of its target genes. These results demonstrate that Exp4 acts as a Sox9 co-regulator that directly regulates binding of Sox9 to its target genes.