Merck
  • Characterization of the Na+, K(+)-ATPase in isolated bovine articular chondrocytes; molecular evidence for multiple alpha and beta isoforms.
Characterization of the Na+, K(+)-ATPase in isolated bovine articular chondrocytes; molecular evidence for multiple alpha and beta isoforms.
Cell biology international (1997-04-01)
A Mobasheri, R J Errington, S Golding, A C Hall, J P Urban
ZUSAMMENFASSUNG
We have used isoform-specific antibodies against the Na+, K(+)-ATPase alpha (alpha 1, alpha 2 and alpha 3) and beta (beta 1 and beta 2) subunit isoforms in order to establish their specific localization in isolated bovine articular chondrocytes. Immunoblotting confirmed the presence of the alpha 1 and alpha 3 isoforms, although alpha 1 expression was significantly greater than alpha 3 as assessed by immunofluorescence confocal laser scanning microscopy and PCR. A similar approach revealed the presence of the beta 1 and beta 2 isoforms in chondrocytes, although beta 2 immunostaining on the plasma membrane was more punctate than beta 1 which in contrast predominated in a subcellular compartment. The plasma membrane abundance of the Na+, K(+)-ATPase was found to be sensitive to the extracellular ionic concentration and long-term elevation of extracellular Na+ concentration significantly upregulated Na+, K(+)-ATPase density as measured by specific 3H-ouabain binding. Our observations suggest that the expression of alpha 3 and beta 2 is not restricted to excitable tissues as previously reported. The physiological relevance of alpha 3 expression in chondrocytes may be related to its low affinity for intracellular Na+ in an extracellular environment where Na+ concentration is unusually high (260-350 mM) compared to other cell types (140 mM). Glycoproteins and their branched carbohydrates have been implicated in cell recognition events, thus the beta 2 subunit glycoprotein may allow the chondrocyte to detect changes in its extracellular environment by physically interacting with components of the cellular cytoskeleton and matrix macromolecules.