Merck
Total synthesis of natural myriaporones.
Angewandte Chemie (International ed. in English) (2004-03-24)
Marta Pérez, Carlos del Pozo, Fernando Reyes, Alberto Rodríguez, Andrés Francesch, Antonio M Echavarren, Carmen Cuevas
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methoxy-N-methylacetamid, 98%