- Novel sulfonate analogues of combretastatin A-4: potent antimitotic agents.
Novel sulfonate analogues of combretastatin A-4: potent antimitotic agents.
Bioorganic & medicinal chemistry letters (2001-04-11)
S L Gwaltney, H M Imade, K J Barr, Q Li, L Gehrke, R B Credo, R B Warner, J Y Lee, P Kovar, J Wang, M A Nukkala, N A Zielinski, D Frost, S C Ng, H L Sham
PMID11294380
ZUSAMMENFASSUNG
Sulfonate analogues of combretastatin A-4 have been prepared. These compounds compete with colchicine and combretastatin A-4 for the colchicine binding site on tubulin and are potent inhibitors of tubulin polymerization and cell proliferation. Importantly, these compounds also inhibit the proliferation of P-glycoprotein positive (+) cancer cells, which are resistant to many other antitumor agents.