Merck
Field resistance of
Journal of pest science (2013-08-24)
Hong Tong, Qi Su, Xiaomao Zhou, Lianyang Bai
ZUSAMMENFASSUNG
The present studies were carried out to evaluate resistance in the populations of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Indoxacarb, PESTANAL®, analytical standard
Supelco
Methomyl, PESTANAL®, analytical standard