Merck
Functional isogenic modeling of BRCA1 alleles reveals distinct carrier phenotypes.
Oncotarget (2015-08-08)
Rory L Cochran, Justin Cidado, Minsoo Kim, Daniel J Zabransky, Sarah Croessmann, David Chu, Hong Yuen Wong, Julia A Beaver, Karen Cravero, Bracha Erlanger, Heather Parsons, Christopher M Heaphy, Alan K Meeker, Josh Lauring, Ben Ho Park
ZUSAMMENFASSUNG
Clinical genetic testing of BRCA1 and BRCA2 is commonly performed to identify specific individuals at risk for breast and ovarian cancers who may benefit from prophylactic therapeutic interventions. Unfortunately, it is evident that deleterious BRCA1 alleles demonstrate variable penetrance and that many BRCA1 variants of unknown significance (VUS) exist. In order to further refine hereditary risks that may be associated with specific BRCA1 alleles, we performed gene targeting to establish an isogenic panel of immortalized human breast epithelial cells harboring eight clinically relevant BRCA1 alleles. Interestingly, BRCA1 mutations and VUS had distinct, quantifiable phenotypes relative to isogenic parental BRCA1 wild type cells and controls. Heterozygous cells with known deleterious BRCA1 mutations (185delAG, C61G and R71G) demonstrated consistent phenotypes in radiation sensitivity and genomic instability assays, but showed variability in other assays. Heterozygous BRCA1 VUS cells also demonstrated assay variability, with some VUS demonstrating phenotypes more consistent with deleterious alleles. Taken together, our data suggest that BRCA1 deleterious mutations and VUS can differ in their range of tested phenotypes, suggesting they might impart varying degrees of risk. These results demonstrate that functional isogenic modeling of BRCA1 alleles could aid in classifying BRCA1 mutations and VUS, and determining BRCA allele cancer risk.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution