Merck
  • Synthesis and biological evaluation of achiral indole-substituted titanocene dichloride derivatives.
Synthesis and biological evaluation of achiral indole-substituted titanocene dichloride derivatives.
International journal of medicinal chemistry (2012-01-01)
Anthony Deally, Frauke Hackenberg, Grainne Lally, Matthias Tacke
ZUSAMMENFASSUNG
Six new titanocene compounds have been isolated and characterised. These compounds were synthesised from their fulvene precursors using Super Hydride (LiBEt3H) followed by transmetallation with titanium tetrachloride to yield the corresponding titanocene dichloride derivatives. These complexes are bis-[((1-methyl-3-diethylaminomethyl)indol-2-yl)methylcyclopentadienyl] titanium (IV) dichloride (5a), bis-[((5-methoxy-1-methyl,3-diethylaminomethyl)indol-2-yl)methylcyclopentadienyl] titanium (IV) dichloride (5b), bis-[((1-methyl,3-diethylaminomethyl)indol-4-yl)methylcyclopentadienyl] titanium (IV) dichloride (5c), bis-[((5-bromo-1-methyl)indol-3-yl)methylcyclopentadienyl] titanium (IV) dichloride (5d), bis-[((5-chloro-1-methyl)indol-3-yl)methylcyclopentadienyl] titanium (IV) dichloride (5e), and bis-[((5-fluoro-1-methyl)indol-3-yl)methylcyclopentadienyl] titanium (IV) dichloride (5f). All six titanocenes 5a-5f were tested for their cytotoxicity through MTT-based in vitro tests on CAKI-1 cell lines using DMSO and Soluphor P as solubilising agents in order to determine their IC50 values. Titanocenes 5a-5f were found to have IC50 values of 10 (±2), 21 (±3), 29 (±4), 140 (±6), and 450 (±10) μM when tested using DMSO.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indol-5-carboxaldehyd, 98%
Sigma-Aldrich
6-Bromindol, 96%