Merck
  • Unlike pancreatic cancer cells pancreatic cancer associated fibroblasts display minimal gene induction after 5-aza-2'-deoxycytidine.
Unlike pancreatic cancer cells pancreatic cancer associated fibroblasts display minimal gene induction after 5-aza-2'-deoxycytidine.
PloS one (2012-09-18)
Jun Yu, Kimberly Walter, Noriyuki Omura, Seung-Mo Hong, Angela Young, Ang Li, Audrey Vincent, Michael Goggins
ZUSAMMENFASSUNG
Cancer associated stromal fibroblasts (CAFs) undergo transcriptional and phenotypic changes that contribute to tumor progression, but the mechanisms responsible for these changes are not well understood. Aberrant DNA methylation is an important cause of transcriptional alterations in cancer cells but it is not known how important DNA methylation alterations are to CAF behavior. We used Affymetrix exon arrays to compare genes induced by the DNA methylation inhibitor 5-aza-dC in cultured pancreatic cancer associated fibroblasts, pancreatic control fibroblasts and pancreatic cancer cell lines. We found that pancreatic CAFs and control pancreatic fibroblasts were less responsive to 5-aza-dC-mediated gene reactivation than pancreatic cancer cells (mean+/-SD of genes induced ≥ 5-fold was 9 ± 10 genes in 10 pancreatic CAF cultures, 17 ± 14 genes in 3 control pancreatic fibroblast cultures, and 134 ± 85 genes in 4 pancreatic cancer cell lines). We examined differentially expressed genes between CAFs and control fibroblasts for candidate methylated genes and identified the disintegrin and metalloprotease, ADAM12 as hypomethylated and overexpressed in pancreatic CAF lines and overexpressed in fibroblasts adjacent to primary pancreatic adenocarcinomas. Compared to pancreatic cancer cells, few genes are reactivated by DNMT1 inhibition in pancreatic CAFs suggesting these cells do not harbor many functionally important alterations in DNA methylation. CAFs may also not be very responsive to therapeutic targeting with DNA methylation inhibitors.