Merck
  • Cyproheptadine enhances the I(K) of mouse cortical neurons through sigma-1 receptor-mediated intracellular signal pathway.
Cyproheptadine enhances the I(K) of mouse cortical neurons through sigma-1 receptor-mediated intracellular signal pathway.
PloS one (2012-07-31)
Yan-Lin He, Chun-Lei Zhang, Xiao-Fei Gao, Jin-Jing Yao, Chang-Long Hu, Yan-Ai Mei
ZUSAMMENFASSUNG
Cyproheptadine (CPH) is a histamine- and serotonin-receptor antagonist, and its effects are observed recently in the modulation of multiple intracellular signals. In this study, we used cortical neurons and HEK-293 cells transfected with Kv2.1 α-subunit to address whether CPH modify neural voltage-gated K(+) channels by a mechanism independent of its serotonergic and histaminergic properties. Our results demonstrate that intracellularly delivered CPH increased the I(K) by reducing the activity of protein kinas A (PKA). Inhibition of G(i) eliminated the CPH-induced effect on both the I(K) and PKA. Blocking of 5-HT-, M-, D(2)-, H(1)- or H(2)-type GPCR receptors with relevant antagonists did not eliminate the CPH-induced effect on the I(K). Antagonists of the sigma-1 receptor, however, blocked the effect of CPH. Moreover, the inhibition of sigma-1 by siRNA knockdown significantly reduced the CPH-induced effect on the I(K). On the contrary, sigma-1 receptor agonist mimicked the effects of CPH on the induction of I(K). A ligand-receptor binding assay indicated that CPH bound to the sigma-1 receptor. Similar effect of CPH were obtained from HEK-293 cells transfected with the α-subunit of Kv2.1. In overall, we reveal for the first time that CPH enhances the I(K) by modulating activity of PKA, and that the associated activation of the sigma-1 receptor/G(i)-protein pathway might be involved. Our findings illustrate an uncharacterized effect of CPH on neuron excitability through the I(K), which is independent of histamine H(1) and serotonin receptors.