Merck
  • Endocrine control of embryonic diapause in the Australian sharpnose shark Rhizoprionodon taylori.
Endocrine control of embryonic diapause in the Australian sharpnose shark Rhizoprionodon taylori.
PloS one (2014-07-06)
Daniela Waltrick, Susan M Jones, Colin A Simpfendorfer, Cynthia A Awruch
ZUSAMMENFASSUNG
The reproductive cycle of the Australian sharpnose shark, Rhizoprionodon taylori, includes a temporary suspension of development at the commencement of embryogenesis termed embryonic diapause. This study investigated levels of 17β-estradiol (E2), testosterone (T) and progesterone (P4) in plasma samples of mature wild female R. taylori captured throughout the reproductive cycle and correlated them with internal morphological changes. Levels of T were elevated through most of the embryonic diapause period, suggesting a role of this hormone in the maintenance of this condition. Increasing plasma T concentrations from late diapause to early active development were associated with a possible role of androgens in the termination of embryonic diapause. As in other elasmobranchs, a concomitant increase of E2 with ovarian follicle size indicated a direct role of this hormone in regulating vitellogenesis, while a peak in P4 suggested this hormone is associated with preovulation and ovulation. Additionally, significant correlations between photoperiod or water temperature and maximum follicular diameter and hepatosomatic index suggest that these abiotic factors may also play a role triggering and regulating the synchrony and timing of reproductive events.