Merck
Expression analysis of BACE2 in brain and peripheral tissues.
The Journal of biological chemistry (2000-04-06)
B D Bennett, S Babu-Khan, R Loeloff, J C Louis, E Curran, M Citron, R Vassar
ZUSAMMENFASSUNG
Beta-site amyloid precursor protein cleaving enzyme (BACE) is a novel transmembrane aspartic protease that possesses all the known characteristics of the beta-secretase involved in Alzheimer's disease (Vassar, R., Bennett, B. D., Babu-Khan, S., Kahn, S., Mendiaz, E. A., Denis, P., Teplow, D. B., Ross, S., Amarante, P., Loeloff, R., Luo, Y., Fisher, S., Fuller, J., Edenson, S., Lile, J., Jarosinski, M. A., Biere, A. L., Curran, E., Burgess, T., Louis, J. -C., Collins, F., Treanor, J., Rogers, G., and Citron, M. (1999) Science 286, 735-741). We have analyzed the sequence and expression pattern of a BACE homolog termed BACE2. BACE and BACE2 are unique among aspartic proteases in that they possess a carboxyl-terminal extension with a predicted transmembrane region and together they define a new family. Northern analysis reveals that BACE2 mRNA is expressed at low levels in most human peripheral tissues and at higher levels in colon, kidney, pancreas, placenta, prostate, stomach, and trachea. Human adult and fetal whole brain and most adult brain subregions express very low or undetectable levels of BACE2 mRNA. In addition, in situ hybridization of adult rat brain shows that BACE2 mRNA is expressed at very low levels in most brain regions. The very low or undetectable levels of BACE2 mRNA in the brain are not consistent with the expression pattern predicted for beta-secretase.