Merck
  • Multifunctional peptides derived from an egg yolk protein hydrolysate: isolation and characterization.
Multifunctional peptides derived from an egg yolk protein hydrolysate: isolation and characterization.
Amino acids (2014-11-20)
Aleksandra Zambrowicz, Marta Pokora, Bartosz Setner, Anna Dąbrowska, Marek Szołtysik, Konrad Babij, Zbigniew Szewczuk, Tadeusz Trziszka, Gert Lubec, Józefa Chrzanowska
ZUSAMMENFASSUNG
An egg yolk protein by-product following ethanol extraction of phospholipids (YP) was hydrolyzed with pepsin to produce and identify novel peptides that revealed antioxidant, ACE inhibitory and antidiabetic (α-glucosidase and DPP-IV inhibitory) activities. The peptic hydrolysate of YP was fractionated by ion-exchange chromatography and reversed-phase high-pressure liquid chromatography. Isolated peptides were identified using mass spectrometry (MALDI-ToF) and the Mascot Search Results database. Four peptides of MW ranging from 1,210.62 to 1,677.88 Da corresponded to the fragments of Apolipoprotein B (YINQMPQKSRE; YINQMPQKSREA), Vitellogenin-2 (VTGRFAGHPAAQ) and Apovitellenin-1 (YIEAVNKVSPRAGQF). These peptides were chemically synthesized and showed antioxidant, ACE inhibitory or/and antidiabetic activities. Peptide YIEAVNKVSPRAGQF exerted the strongest ACE inhibitory activity, with IC50 = 9.4 µg/mL. The peptide YINQMPQKSRE showed the strongest DPPH free radical scavenging and DPP-IV inhibitory activities and its ACE inhibitory activity (IC50) reached 10.1 µg/mL. The peptide VTGRFAGHPAAQ revealed the highest α-glucosidase inhibitory activity (IC50 = 365.4 µg/mL). A novel nutraceutical effect for peptides from an egg yolk hydrolysate was shown.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazin-p,p′-disulfonsäure Mononatriumsalz Hydrat, 97%
Sigma-Aldrich
α-Glucosidase aus Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein