Merck
The Plasmodium PHIST and RESA-Like Protein Families of Human and Rodent Malaria Parasites.
PloS one (2016-03-31)
Cristina K Moreira, Bernina Naissant, Alida Coppi, Brandy L Bennett, Elena Aime, Blandine Franke-Fayard, Chris J Janse, Isabelle Coppens, Photini Sinnis, Thomas J Templeton
ZUSAMMENFASSUNG
The phist gene family has members identified across the Plasmodium genus, defined by the presence of a domain of roughly 150 amino acids having conserved aromatic residues and an all alpha-helical structure. The family is highly amplified in P. falciparum, with 65 predicted genes in the genome of the 3D7 isolate. In contrast, in the rodent malaria parasite P. berghei 3 genes are identified, one of which is an apparent pseudogene. Transcripts of the P. berghei phist genes are predominant in schizonts, whereas in P. falciparum transcript profiles span different asexual blood stages and gametocytes. We pursued targeted disruption of P. berghei phist genes in order to characterize a simplistic model for the expanded phist gene repertoire in P. falciparum. Unsuccessful attempts to disrupt P. berghei PBANKA_114540 suggest that this phist gene is essential, while knockout of phist PBANKA_122900 shows an apparent normal progression and non-essential function throughout the life cycle. Epitope-tagging of P. falciparum and P. berghei phist genes confirmed protein export to the erythrocyte cytoplasm and localization with a punctate pattern. Three P. berghei PEXEL/HT-positive exported proteins exhibit at least partial co-localization, in support of a common vesicular compartment in the cytoplasm of erythrocytes infected with rodent malaria parasites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-c-Myc, clone 9E10, purified immunoglobulin, buffered aqueous solution