Merck
A trial of somatic gene targeting in vivo with an adenovirus vector.
Genetic vaccines and therapy (2005-10-13)
Asami Ino, Yasuhiro Naito, Hiroyuki Mizuguchi, Naofumi Handa, Takao Hayakawa, Ichizo Kobayashi
ZUSAMMENFASSUNG
Gene targeting in vivo provides a potentially powerful method for gene analysis and gene therapy. In order to sensitively detect and accurately measure designed sequence changes, we have used a transgenic mouse system, MutaMouse, which has been developed for detection of mutation in vivo. It carries bacteriophage lambda genome with lacZ+ gene, whose change to lacZ-negative allele is detected after in vitro packaging into bacteriophage particles. We have also demonstrated that gene transfer with a replication-defective adenovirus vector can achieve efficient and accurate gene targeting in vitro. An 8 kb long DNA corresponding to the bacteriophage lambda transgene with one of two lacZ-negative single-base-pair-substitution mutant allele was inserted into a replication-defective adenovirus vector. This recombinant adenovirus was injected to the transgenic mice via tail-vein. Twenty-four hours later, genomic DNA was extracted from the liver tissue and the lambda::lacZ were recovered by in vitro packaging. The lacZ-negative phage was detected as a plaque former on agar with phenyl-beta-D-galactoside. The mutant frequency of the lacZ-negative recombinant adenovirus injected mice was at the same level with the control mouse (approximately 1/10000). Our further restriction analysis did not detect any designed recombinant. The frequency of gene targeting in the mouse liver by these recombinant adenoviruses was shown to be less than 1/20000 in our assay. However, these results will aid the development of a sensitive, reliable and PCR-independent assay for gene targeting in vivo mediated by virus vectors and other means.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenyl-β-D-Galaktopyranosid, ≥98% (TLC)