Merck
  • The sphingosine-1-phosphate (S1P) lysophospholipid receptor S1P3 regulates MAdCAM-1+ endothelial cells in splenic marginal sinus organization.
The sphingosine-1-phosphate (S1P) lysophospholipid receptor S1P3 regulates MAdCAM-1+ endothelial cells in splenic marginal sinus organization.
The Journal of experimental medicine (2004-12-08)
Irute Girkontaite, Vadim Sakk, Martin Wagner, Tilman Borggrefe, Kerry Tedford, Jerold Chun, Klaus-Dieter Fischer
ZUSAMMENFASSUNG
Marginal zones (MZs) are microdomains in the spleen that contain various types of immune cells, including MZ B cells, MOMA1(+) metallophilic macrophages, and mucosal addressin cell adhesion molecule 1 (MAdCAM-1)(+) endothelial cells. MAdCAM-1(+) and MOMA1(+) cells line the sinus, that separates MZs from splenic follicles. Here we show that a receptor for the lysophospholipid sphingosine-1-phosphate (S1P), S1P(3), is required for normal numbers of splenic immature and MZ B cells, and for S1P-induced chemotaxis of MZ B cells. S1P(3) is also essential for proper alignment of MOMA1(+) macrophages and MAdCAM-1(+) endothelial cells along the marginal sinus. The lack of cohesion of the marginal sinus in S1P(3)(-/-) mice affects MZ B cell functions, as wild-type (WT) MZ B cells migrate more into S1P(3)(-/-) follicles than into WT follicles after treatment with lipopolysaccharide. Additionally, short-term homing experiments demonstrate that WT MZ B cells home to the S1P(3)(-/-) spleen in increased numbers, suggesting a role for the marginal sinus in regulating MZ B cells numbers. Moreover, S1P(3)(-/-) mice are defective in mounting immune responses to thymus-independent antigen type 2 due to defects in radiation-resistant cells in the spleen. These data identify lysophospholipids and the S1P(3) receptor as essential regulators of the MZ sinus and its role as a barrier to the follicle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
ChemiSCREEN Membrane Preparation Recombinant Human S1P3 Lysophospholipid Receptor, Human S1P3 / EDG3 GPCR membrane preparation for GTPγS Binding & Radioligand Binding Assays.