Merck
  • Cryptotanshinone Reverses Reproductive and Metabolic Disturbances in PCOS Model Rats via Regulating the Expression of CYP17 and AR.
Cryptotanshinone Reverses Reproductive and Metabolic Disturbances in PCOS Model Rats via Regulating the Expression of CYP17 and AR.
Evidence-based complementary and alternative medicine : eCAM (2014-05-07)
Jin Yu, Dongxia Zhai, Li Hao, Danying Zhang, Lingling Bai, Zailong Cai, Chaoqin Yu
ZUSAMMENFASSUNG
Objective. To explore the effect of Cryptotanshinone on reversing the reproductive and metabolic disturbances in polycystic ovary syndrome (PCOS) model rats and the possible regulatory mechanisms. Methods. PCOS model rats were induced by subcutaneous injection of dehydroepiandrosterone (DHEA) and verified by histological screening of vaginal exfoliated cells. After Cryptotanshinone intervention, the rats' body weight and ovary morphological were observed; the serum biochemical assessments were analyzed by radioimmunoassay (RIA) and key genes and proteins related with anabolism of androgen and insulin were detected by Real-Time PCR and Immunohistochemical (IHC). Results. The estrous cyclicity of PCOS model rats was significantly recovered by Cryptotanshinone. The body weight, ovarian coefficient, and ovarian morphology had been improved and the serum biochemical indicators including testosterone (T), androstenedione (A2), luteinizing hormone (LH), LH/follicle stimulating hormone (FSH), sexual binding globulin (SHBG), low density cholesterol (LDL-C), fasting insulin (FINS) were reversed after Cryptotanshinone intervention. Specifically, the levels of Cytochrome P450, 17-a hydroxylase/17,20 lyase (CYP17), and androgen receptor (AR) were downregulated significantly. Conclusions. Our data suggest that Cryptotanshinone could rebalance reproductive and metabolic disturbances in PCOS model rats and could be a potential therapeutic agent for the treatment of PCOS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sesamöl, Antioxidant, delivery vehicle for fat-soluble compounds