Merck
  • Enhanced expression of glucose-regulated protein 78 correlates with malondialdehyde levels during the formation of liver cirrhosis in rats.
Enhanced expression of glucose-regulated protein 78 correlates with malondialdehyde levels during the formation of liver cirrhosis in rats.
Experimental and therapeutic medicine (2015-12-17)
Yun Zhang, Huiying Zhang, Zhongfu Zhao, Minli Lv, Jiantao Jia, Lili Zhang, Xiaoxia Tian, Yunxia Chen, Baohong Li, Mingshe Liu, Dewu Han, Cheng Ji
ZUSAMMENFASSUNG
The aim of the present study was to explore the role of glucose-regulated protein 78 (GRP78) in the development of liver cirrhosis promoted by intestinal endotoxemia in rats. Fifty-one male Wistar rats were randomly divided into the liver cirrhosis 4-week, 6-week and 8-week groups and the normal control group at each time point. Liver cirrhosis was induced by employing multiple pathogenic factors in the rats. Blood and liver tissues were collected. The levels of alanine aminotransferase (ALT), homocysteine, endotoxin and tumor necrosis factor-α (TNF-α) in the plasma, and TNF-α, malondialdehyde (MDA) and procollagen type III peptide (PIIIP) in the liver tissues were determined. The mRNA and protein expression levels of GRP78 in the liver were detected using reverse transcription-quantitative polymerase chain reaction and immunohistochemistry. Morphological changes were observed through hematoxylin and eosin and van Gieson staining of the liver. Liver cirrhosis caused marked histopathological changes to the livers of the rats. Following significant increases in the levels of ALT, homocysteine, endotoxin and TNF-α in the plasma, and TNF-α, MDA and PIIIP in the liver tissues of all experimental groups with the progression of liver cirrhosis, the mRNA and protein expression levels of GRP78 also gradually increased. In addition, correlation analysis indicated that the enhanced expression of GRP78 correlated with the MDA levels of the rats during the formation of liver cirrhosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GRP78/BiP (ET-21) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution