Merck
  • THOC5 controls 3'end-processing of immediate early genes via interaction with polyadenylation specific factor 100 (CPSF100).
THOC5 controls 3'end-processing of immediate early genes via interaction with polyadenylation specific factor 100 (CPSF100).
Nucleic acids research (2014-10-03)
Doan Duy Hai Tran, Shashank Saran, Andrew J K Williamson, Andrew Pierce, Oliver Dittrich-Breiholz, Lutz Wiehlmann, Alexandra Koch, Anthony D Whetton, Teruko Tamura
ZUSAMMENFASSUNG
Transcription of immediate early genes (IEGs) in response to extrinsic and intrinsic signals is tightly regulated at multiple stages. It is known that untranslated regions of the RNA can play a role in these processes. Here we show that THOC5, a member of the TREX (transcription/export) complex, plays a role in expression of only a subset of constitutively active genes, however transcriptome analysis reveals that more than 90% of IEG were not induced by serum in THOC5 depleted cells. Furthermore, THOC5 depletion does not influence the expression of the most rapidly induced IEGs, e.g. Fos and Jun. One group of THOC5 target genes, including Id1, Id3 and Wnt11 transcripts, were not released from chromatin in THOC5 depleted cells. Genes in another group, including Myc and Smad7 transcripts, were released with shortening of 3'UTR by alternative cleavage, and were spliced but export was impaired in THOC5 depleted cells. By interactome analysis using THOC5 as bait, we show that upon stimulation with serum THOC5 forms a complex with polyadenylation-specific factor 100 (CPSF100). THOC5 is required for recruitment of CPSF100 to 3'UTR of THOC5 target genes. These data suggest the presence of a novel mechanism for the control of IEG response by THOC5 via 3'end-processing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human CPSF2