- Rac1 and a GTPase-activating protein, MgcRacGAP, are required for nuclear translocation of STAT transcription factors.
Rac1 and a GTPase-activating protein, MgcRacGAP, are required for nuclear translocation of STAT transcription factors.
The Journal of cell biology (2006-12-21)
Toshiyuki Kawashima, Ying Chun Bao, Yasushi Nomura, Yuseok Moon, Yukio Tonozuka, Yukinori Minoshima, Tomonori Hatori, Akiho Tsuchiya, Mari Kiyono, Tetsuya Nosaka, Hideaki Nakajima, David A Williams, Toshio Kitamura
PMID17178910
ZUSAMMENFASSUNG
STAT transcription factors are tyrosine phosphorylated upon cytokine stimulation and enter the nucleus to activate target genes. We show that Rac1 and a GTPase-activating protein, MgcRacGAP, bind directly to p-STAT5A and are required to promote its nuclear translocation. Using permeabilized cells, we find that nuclear translocation of purified p-STAT5A is dependent on the addition of GTP-bound Rac1, MgcRacGAP, importin alpha, and importin beta. p-STAT3 also enters the nucleus via this transport machinery, and mutant STATs lacking the MgcRacGAP binding site do not enter the nucleus even after phosphorylation. We conclude that GTP-bound Rac1 and MgcRacGAP function as a nuclear transport chaperone for activated STATs.
MATERIALIEN