Merck
  • A Comparison of Neutral Mg2+-Selective Ionophores in Solvents Polymeric Membranes: Complex Stoichiometry
A Comparison of Neutral Mg<SUP>2+</SUP>-Selective Ionophores in Solvents Polymeric Membranes: Complex Stoichiometry
Analytical Sciences (2000)
Zhang, W., et al.