Merck
  • Milligram production and biological activity characterization of the human chemokine receptor CCR3.
Milligram production and biological activity characterization of the human chemokine receptor CCR3.
PloS one (2013-06-12)
Mingqing Wang, Baosheng Ge, Renmin Li, Xiaoqiang Wang, Jun Lao, Fang Huang
ZUSAMMENFASSUNG
Human chemokine receptor CCR3 (hCCR3) belongs to the G protein-coupled receptors (GPCRs) superfamily of membrane proteins and plays major roles in allergic diseases and angiogenesis. In order to study the structural and functional mechanism of hCCR3, it is essential to produce pure protein with biological functions on a milligram scale. Here we report the expression of hCCR3 gene in a tetracycline-inducible stable mammalian cell line. A cell clone with high hCCR3 expression was selected from 46 stably transfected cell clones and from this cell line pure hCCR3 on a milligram scale was obtained after two-step purification. Circular dichroism spectrum with a characteristic shape and magnitude for α-helix indicated proper folding of hCCR3 after purification. The biological activity of purified hCCR3 was verified by its high binding affinity with its endogenous ligands CCL11 and CCL24, with K D in the range of 10(-8) M to 10(-6) M.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterylhemisuccinat Tris-salz, anionic detergent