Merck
BET bromodomain inhibition suppresses TH17-mediated pathology.
The Journal of experimental medicine (2013-10-09)
Deanna A Mele, Andres Salmeron, Srimoyee Ghosh, Hon-Ren Huang, Barbara M Bryant, Jose M Lora
ZUSAMMENFASSUNG
Interleukin (IL) 17-producing T helper (T(H)17) cells have been selected through evolution for their ability to control fungal and bacterial infections. It is also firmly established that their aberrant generation and activation results in autoimmune conditions. Using a characterized potent and selective small molecule inhibitor, we show that the bromodomain and extra-terminal domain (BET) family of chromatin adaptors plays fundamental and selective roles in human and murine T(H)17 differentiation from naive CD4(+) T cells, as well as in the activation of previously differentiated T(H)17 cells. We provide evidence that BET controls T(H)17 differentiation in a bromodomain-dependent manner through a mechanism that includes the direct regulation of multiple effector T(H)17-associated cytokines, including IL17, IL21, and GMCSF. We also demonstrate that BET family members Brd2 and Brd4 associate with the Il17 locus in T(H)17 cells, and that this association requires bromodomains. We recapitulate the critical role of BET bromodomains in T(H)17 differentiation in vivo and show that therapeutic dosing of the BET inhibitor is efficacious in mouse models of autoimmunity. Our results identify the BET family of proteins as a fundamental link between chromatin signaling and T(H)17 biology, and support the notion of BET inhibition as a point of therapeutic intervention in autoimmune conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
MILLIPLEX® Human Th17 Panel, Configurable Human Th17 25-Plex Panel