Merck
  • Disruption of microtubules sensitizes the DNA damage-induced apoptosis through inhibiting nuclear factor κB (NF-κB) DNA-binding activity.
Disruption of microtubules sensitizes the DNA damage-induced apoptosis through inhibiting nuclear factor κB (NF-κB) DNA-binding activity.
Journal of Korean medical science (2010-11-10)
Hyunji Lee, Juhee Jeon, Young Sue Ryu, Jae Eun Jeong, Sanghee Shin, Tiejun Zhang, Seong Wook Kang, Jang Hee Hong, Gang Min Hur
ZUSAMMENFASSUNG
The massive reorganization of microtubule network involves in transcriptional regulation of several genes by controlling transcriptional factor, nuclear factor-kappa B (NF-κB) activity. The exact molecular mechanism by which microtubule rearrangement leads to NF-κB activation largely remains to be identified. However microtubule disrupting agents may possibly act in synergy or antagonism against apoptotic cell death in response to conventional chemotherapy targeting DNA damage such as adriamycin or comptothecin in cancer cells. Interestingly pretreatment of microtubule disrupting agents (colchicine, vinblastine and nocodazole) was observed to lead to paradoxical suppression of DNA damage-induced NF-κB binding activity, even though these could enhance NF-κB signaling in the absence of other stimuli. Moreover this suppressed NF-κB binding activity subsequently resulted in synergic apoptotic response, as evident by the combination with Adr and low doses of microtubule disrupting agents was able to potentiate the cytotoxic action through caspase-dependent pathway. Taken together, these results suggested that inhibition of microtubule network chemosensitizes the cancer cells to die by apoptosis through suppressing NF-κB DNA binding activity. Therefore, our study provided a possible anti-cancer mechanism of microtubule disrupting agent to overcome resistance against to chemotherapy such as DNA damaging agent.