Merck
  • Ectopic over-expression of tristetraprolin in human cancer cells promotes biogenesis of let-7 by down-regulation of Lin28.
Ectopic over-expression of tristetraprolin in human cancer cells promotes biogenesis of let-7 by down-regulation of Lin28.
Nucleic acids research (2012-01-03)
Chae Won Kim, Mai-Tram Vo, Hong Kyeung Kim, Hyun Hee Lee, Nal Ae Yoon, Byung Ju Lee, Young Joo Min, Won Duk Joo, Hee Jeong Cha, Jeong Woo Park, Wha Ja Cho
ZUSAMMENFASSUNG
Tristetraprolin (TTP) is a AU-rich element (ARE) binding protein and exhibits suppressive effects on cell growth through down-regulation of ARE-containing oncogenes. The let-7 microRNA has emerged as a significant factor in tumor suppression. Both TTP and let-7 are often repressed in human cancers, thereby promoting oncogenesis by derepressing their target genes. In this work, an unexpected link between TTP and let-7 has been found in human cancer cells. TTP promotes an increase in expression of mature let-7, which leads to the inhibition of let-7 target gene CDC34 expression and suppresses cell growth. This event is associated with TTP-mediated inhibition of Lin28, which has emerged as a negative modulator of let-7. Lin28 mRNA contains ARE within its 3'-UTR and TTP enhances the decay of Lin28 mRNA through binding to its 3'-UTR. This suggests that the TTP-mediated down-regulation of Lin28 plays a key role in let-7 miRNA biogenesis in cancer cells.