Merck
  • Human tumor necrosis factor (TNF)-alpha-induced protein 8-like 2 suppresses hepatocellular carcinoma metastasis through inhibiting Rac1.
Human tumor necrosis factor (TNF)-alpha-induced protein 8-like 2 suppresses hepatocellular carcinoma metastasis through inhibiting Rac1.
Molecular cancer (2013-11-28)
Xuelei Cao, Li Zhang, Yongyu Shi, Yue Sun, Shen Dai, Chun Guo, Faliang Zhu, Qun Wang, Jianing Wang, Xiaoyan Wang, Youhai H Chen, Lining Zhang
ZUSAMMENFASSUNG
Tumor invasion and metastasis are the major reasons for leading death of patients with hepatocellular carcinoma (HCC). Therefore, to identify molecules that can suppress invasion and metastasis of tumor will provide novel targets for HCC therapies. Tumor necrosis factor (TNF)-alpha-induced protein 8-like 2, TIPE2, is a novel immune negative molecule and an inhibitor of the oncogenic Ras in mice but its function in human is unclear. Our previous research has shown that TIPE2 is downregulated in human primary HCC compared with the paired adjacent non-tumor tissues. In present study, we provide evidence that TIPE2 inhibits effectively human hepatocellular carcinoma metastasis. The forced expression of TIPE2 in HCC-derived cell lines markedly inhibits tumor cell growth, migration and invasion in vitro and suppresses growth and metastasis of HCC in vivo. Clinical information from a cohort of 112 patients reveals that loss or reduced expression of TIPE2 in primary HCC tissues is significantly associated with tumor metastasis. Mechanically, TIPE2 inhibits the migration and invasion through targeting Rac1 and then reduces F-actin polymerization and expression of matrix metallopeptidase 9 (MMP9) and urokinase plasminogen activator (uPA). Our results indicate that human TIPE2 is endogenous inhibitor of Rac1 in HCC by which it attenuates invasion and metastasis of HCC. The data suggest that TIPE2 will be a new target for HCC therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFAIP8L2