Merck
  • Dicer1 ablation in the mouse epididymis causes dedifferentiation of the epithelium and imbalance in sex steroid signaling.
Dicer1 ablation in the mouse epididymis causes dedifferentiation of the epithelium and imbalance in sex steroid signaling.
PloS one (2012-06-16)
Ida Björkgren, Lauri Saastamoinen, Anton Krutskikh, Ilpo Huhtaniemi, Matti Poutanen, Petra Sipilä
ZUSAMMENFASSUNG
The postnatal development of the epididymis is a complex process that results in a highly differentiated epithelium, divided into several segments. Recent studies indicate a role for RNA interference (RNAi) in the development of the epididymis, however, the actual requirement for RNAi has remained elusive. Here, we present the first evidence of a direct need for RNAi in the differentiation of the epididymal epithelium. By utilizing the Cre-LoxP system we have generated a conditional knock-out of Dicer1 in the two most proximal segments of the mouse epididymis. Recombination of Dicer1, catalyzed by Defb41(iCre/wt), took place before puberty, starting from 12 days postpartum. Shortly thereafter, downregulation of the expression of two genes specific for the most proximal epididymis (lipocalin 8 and cystatin 8) was observed. Following this, segment development continued until week 5 at which age the epithelium started to regress back to an undifferentiated state. The dedifferentiated epithelium also showed an increase in estrogen receptor 1 expression while the expression of androgen receptor and its target genes; glutathione peroxidase 5, lipocalin 5 and cysteine-rich secretory protein 1 was downregulated, indicating imbalanced sex steroid signaling. At the time of the final epididymal development, Dicer1 acts as a regulator of signaling pathways essential for maintaining epithelial cell differentiation.