Merck
  • Dopamine signaling negatively regulates striatal phosphorylation of Cdk5 at tyrosine 15 in mice.
Dopamine signaling negatively regulates striatal phosphorylation of Cdk5 at tyrosine 15 in mice.
Frontiers in cellular neuroscience (2013-02-20)
Yukio Yamamura, Ryoma Morigaki, Jiro Kasahara, Hironori Yokoyama, Akie Tanabe, Shinya Okita, Hidetaka Koizumi, Shinji Nagahiro, Ryuji Kaji, Satoshi Goto
ZUSAMMENFASSUNG
Striatal functions depend on the activity balance between the dopamine and glutamate neurotransmissions. Glutamate inputs activate cyclin-dependent kinase 5 (Cdk5), which inhibits postsynaptic dopamine signaling by phosphorylating DARPP-32 (dopamine- and cAMP-regulated phosphoprotein, 32 kDa) at Thr75 in the striatum. c-Abelson tyrosine kinase (c-Abl) is known to phosphorylate Cdk5 at Tyr15 (Tyr15-Cdk5) and thereby facilitates the Cdk5 activity. We here report that Cdk5 with Tyr15 phosphorylation (Cdk5-pTyr15) is enriched in the mouse striatum, where dopaminergic stimulation inhibited phosphorylation of Tyr15-Cdk5 by acting through the D2 class dopamine receptors. Moreover, in the 1-methyl-4-phenyl-1,2,4,6-tetrahydropyridine (MPTP) mouse model, dopamine deficiency caused increased phosphorylation of both Tyr15-Cdk5 and Thr75-DARPP-32 in the striatum, which could be attenuated by administration of L-3,4-dihydroxyphenylalanine and imatinib (STI-571), a selective c-Abl inhibitor. Our results suggest a functional link of Cdk5-pTyr15 with postsynaptic dopamine and glutamate signals through the c-Abl kinase activity in the striatum.