Merck
  • A universal system for highly efficient cardiac differentiation of human induced pluripotent stem cells that eliminates interline variability.
A universal system for highly efficient cardiac differentiation of human induced pluripotent stem cells that eliminates interline variability.
PloS one (2011-04-16)
Paul W Burridge, Susan Thompson, Michal A Millrod, Seth Weinberg, Xuan Yuan, Ann Peters, Vasiliki Mahairaki, Vassilis E Koliatsos, Leslie Tung, Elias T Zambidis
ZUSAMMENFASSUNG
The production of cardiomyocytes from human induced pluripotent stem cells (hiPSC) holds great promise for patient-specific cardiotoxicity drug testing, disease modeling, and cardiac regeneration. However, existing protocols for the differentiation of hiPSC to the cardiac lineage are inefficient and highly variable. We describe a highly efficient system for differentiation of human embryonic stem cells (hESC) and hiPSC to the cardiac lineage. This system eliminated the variability in cardiac differentiation capacity of a variety of human pluripotent stem cells (hPSC), including hiPSC generated from CD34(+) cord blood using non-viral, non-integrating methods. We systematically and rigorously optimized >45 experimental variables to develop a universal cardiac differentiation system that produced contracting human embryoid bodies (hEB) with an improved efficiency of 94.7±2.4% in an accelerated nine days from four hESC and seven hiPSC lines tested, including hiPSC derived from neonatal CD34(+) cord blood and adult fibroblasts using non-integrating episomal plasmids. This cost-effective differentiation method employed forced aggregation hEB formation in a chemically defined medium, along with staged exposure to physiological (5%) oxygen, and optimized concentrations of mesodermal morphogens BMP4 and FGF2, polyvinyl alcohol, serum, and insulin. The contracting hEB derived using these methods were composed of high percentages (64-89%) of cardiac troponin I(+) cells that displayed ultrastructural properties of functional cardiomyocytes and uniform electrophysiological profiles responsive to cardioactive drugs. This efficient and cost-effective universal system for cardiac differentiation of hiPSC allows a potentially unlimited production of functional cardiomyocytes suitable for application to hPSC-based drug development, cardiac disease modeling, and the future generation of clinically-safe nonviral human cardiac cells for regenerative medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(vinylalkohol), 87-90% hydrolyzed, average mol wt 30,000-70,000