Merck
  • Somatostatin and opioid receptors do not regulate proliferation or apoptosis of the human multiple myeloma U266 cells.
Somatostatin and opioid receptors do not regulate proliferation or apoptosis of the human multiple myeloma U266 cells.
Journal of experimental & clinical cancer research : CR (2009-06-09)
Céline Kerros, Thibault Cavey, Brigitte Sola, Philippe Jauzac, Stéphane Allouche
ZUSAMMENFASSUNG
opioid and somatostatin receptors (SSTRs) that can assemble as heterodimer were individually reported to modulate malignant cell proliferation and to favour apoptosis. SSTRs and opioid receptors expression were examined by RT-PCR, western-blot and binding assays, cell proliferation was studied by XTT assay and propidium iodide (PI) staining and apoptosis by annexin V-PI labelling. almost all human malignant haematological cell lines studied here expressed the five SSTRs. Further experiments were conducted on the human U266 multiple myeloma cells, which express also micro-opioid receptors (MOP-R). XTT assays and cell cycle studies provide no evidence for a significant effect upon opioid or somatostatin receptors stimulation. Furthermore, neither direct effect nor potentiation of the Fas-receptor pathway was detected on apoptosis after these treatments. these data suggest that SSTRs or opioid receptors expression is not a guaranty for an anti-tumoral action in U266 cell line.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Octreotide, ≥98% (HPLC)