Merck
The Fanconi anemia group C protein interacts with uncoordinated 5A and delays apoptosis.
PloS one (2014-03-29)
FengFei Huang, Manel Ben Aissa, Audrey Magron, Caroline C Huard, Chantal Godin, Georges Lévesque, Madeleine Carreau
ZUSAMMENFASSUNG
The Fanconi anemia group C protein (FANCC) is one of the several proteins that comprise the Fanconi anemia (FA) network involved in genomic surveillance. FANCC is mainly cytoplasmic and has many functions, including apoptosis suppression through caspase-mediated proteolytic processing. Here, we examined the role of FANCC proteolytic fragments by identifying their binding partners. We performed a yeast two-hybrid screen with caspase-mediated FANCC cleavage products and identified the dependence receptor uncoordinated-5A (UNC5A) protein. Here, we show that FANCC physically interacts with UNC5A, a pro-apoptotic dependence receptor. FANCC interaction occurs through the UNC5A intracellular domain, specifically via its death domain. FANCC modulates cell sensitivity to UNC5A-mediated apoptosis; we observed reduced UNC5A-mediated apoptosis in the presence of FANCC and increased apoptosis in FANCC-depleted cells. Our results show that FANCC interferes with UNC5A's functions in apoptosis and suggest that FANCC may participate in developmental processes through association with the dependence receptor UNC5A.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human FANCE