- Analysis of TMEM174 gene expression in various renal cancer types by RNA
Analysis of TMEM174 gene expression in various renal cancer types by RNA
Oncology letters (2014-09-10)
Xiujun Zhang, Fen Hu, Lijun Meng, Lixia Gou, Mengmeng Luo
PMID25202393
ZUSAMMENFASSUNG
Transmembrane protein 174 (TMEM174) mRNA is easily detectable in human kidney tissues and activates AP-1 and promotes 293T cell proliferation. In the present study, RNA