Merck
  • Hope for restoration of dead valuable bulls through cloning using donor somatic cells isolated from cryopreserved semen.
Hope for restoration of dead valuable bulls through cloning using donor somatic cells isolated from cryopreserved semen.
PloS one (2014-03-13)
Naresh L Selokar, Monika Saini, Prabhat Palta, Manmohan S Chauhan, Radheysham Manik, Suresh K Singla
ZUSAMMENFASSUNG
Somatic cells were isolated from cryopreserved semen of 4 buffalo bulls, 3 of which had died over 10 years earlier, and were established in culture. The cells expressed cytokeratin-18, keratin and vimentin indicating that they were of epithelial origin. The cells were used as nuclear donors for hand-made cloning for producing buffalo embryos. The blastocyst rate and quality, as indicated by apoptotic index, were comparable among embryos produced using cells obtained from fresh or frozen-thawed semen or those obtained from conventional cell sources such as skin. Examination of the epigenetic status revealed that the global level of H3K27me3 but not that of H3K9/14ac and H4K5ac differed significantly (P<0.05) among cloned embryos from different bulls. The relative mRNA abundance of HDAC1, DNMT1, P53 and CASPASE 3 but not that of DNMT3a differed in cells and in cloned embryos. Following transfer of 24 cloned embryos produced from fresh semen-derived cells to 12 recipients, one calf weighing 55 kg, which is now 6 months of age and is normal, was born through normal parturition. Following transfer of 20 embryos produced from frozen-thawed semen-derived cells to 10 recipients, 2 became pregnant, one of which aborted in the first trimester; the calf born was severely underweight (17 kg), and died 12 h after birth. The ability of cells derived from fresh and frozen-thawed semen to produce live offspring confirms the ability of these cells to be reprogrammed. Our findings pave the way for restoration of highly precious progeny-tested bulls, which has immense economic importance, and can also be used for restoration of endangered species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonales Anti-Vimentin in Maus hergestellte Antikörper, clone V9, ascites fluid
Sigma-Aldrich
Anti-Keratin-Epithel-Antikörper, Klon AE3, clone AE3, Chemicon®, from mouse