Merck
  • Sustained Pax6 Expression Generates Primate-like Basal Radial Glia in Developing Mouse Neocortex.
Sustained Pax6 Expression Generates Primate-like Basal Radial Glia in Developing Mouse Neocortex.
PLoS biology (2015-08-08)
Fong Kuan Wong, Ji-Feng Fei, Felipe Mora-Bermúdez, Elena Taverna, Christiane Haffner, Jun Fu, Konstantinos Anastassiadis, A Francis Stewart, Wieland B Huttner
ZUSAMMENFASSUNG
The evolutionary expansion of the neocortex in mammals has been linked to enlargement of the subventricular zone (SVZ) and increased proliferative capacity of basal progenitors (BPs), notably basal radial glia (bRG). The transcription factor Pax6 is known to be highly expressed in primate, but not mouse, BPs. Here, we demonstrate that sustaining Pax6 expression selectively in BP-genic apical radial glia (aRG) and their BP progeny of embryonic mouse neocortex suffices to induce primate-like progenitor behaviour. Specifically, we conditionally expressed Pax6 by in utero electroporation using a novel, Tis21-CreERT2 mouse line. This expression altered aRG cleavage plane orientation to promote bRG generation, increased cell-cycle re-entry of BPs, and ultimately increased upper-layer neuron production. Upper-layer neuron production was also increased in double-transgenic mouse embryos with sustained Pax6 expression in the neurogenic lineage. Strikingly, increased BPs existed not only in the SVZ but also in the intermediate zone of the neocortex of these double-transgenic mouse embryos. In mutant mouse embryos lacking functional Pax6, the proportion of bRG among BPs was reduced. Our data identify specific Pax6 effects in BPs and imply that sustaining this Pax6 function in BPs could be a key aspect of SVZ enlargement and, consequently, the evolutionary expansion of the neocortex.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Phospho-Histon H3 (Ser10)-Antikörper, Mitosemarker, Upstate®, from rabbit
Sigma-Aldrich
Anti-γ-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Tubulin III-Antikörper (neuronal), monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PCNA-Antikörper, Klon PC10, clone PC10, Chemicon®, from mouse