Merck
  • Ca2+ release in muscle fibers expressing R4892W and G4896V type 1 ryanodine receptor disease mutants.
Ca2+ release in muscle fibers expressing R4892W and G4896V type 1 ryanodine receptor disease mutants.
PloS one (2013-01-12)
Romain Lefebvre, Claude Legrand, Linda Groom, Robert T Dirksen, Vincent Jacquemond
ZUSAMMENFASSUNG
The large and rapidly increasing number of potentially pathological mutants in the type 1 ryanodine receptor (RyR1) prompts the need to characterize their effects on voltage-activated sarcoplasmic reticulum (SR) Ca(2+) release in skeletal muscle. Here we evaluated the function of the R4892W and G4896V RyR1 mutants, both associated with central core disease (CCD) in humans, in myotubes and in adult muscle fibers. For both mutants expressed in RyR1-null (dyspedic) myotubes, voltage-gated Ca(2+) release was absent following homotypic expression and only partially restored following heterotypic expression with wild-type (WT) RyR1. In muscle fibers from adult WT mice, both mutants were expressed in restricted regions of the fibers with a pattern consistent with triadic localization. Voltage-clamp-activated confocal Ca(2+) signals showed that fiber regions endowed with G4896V-RyR1s exhibited an ∼30% reduction in the peak rate of SR Ca(2+) release, with no significant change in SR Ca(2+) content. Immunostaining revealed no associated change in the expression of either α1S subunit (Cav1.1) of the dihydropyridine receptor (DHPR) or type 1 sarco(endo)plasmic reticulum Ca(2+) ATPase (SERCA1), indicating that the reduced Ca(2+) release resulted from defective RyR1 function. Interestingly, in spite of robust localized junctional expression, the R4892W mutant did not affect SR Ca(2+) release in adult muscle fibers, consistent with a low functional penetrance of this particular CCD-associated mutant.