Merck
  • On the mechanism of [Rh(CO)2Cl]2-catalyzed intermolecular (5 + 2) reactions between vinylcyclopropanes and alkynes.
On the mechanism of [Rh(CO)2Cl]2-catalyzed intermolecular (5 + 2) reactions between vinylcyclopropanes and alkynes.
Journal of the American Chemical Society (2004-07-30)
Zhi-Xiang Yu, Paul A Wender, K N Houk
ZUSAMMENFASSUNG
DFT calculations have been applied to investigate the reaction mechanism of rhodium dimer, [Rh(CO)2Cl]2, catalyzed intermolecular (5 + 2) reactions between vinylcyclopropanes and alkynes. The catalytic species is Rh(CO)Cl and the catalytic cycle is through the sequential reactions of cyclopropyl cleavage of vinylcyclopropane, alkyne insertion (rate-determining step), and a migratory reductive elimination.