Merck
  • TRAF4 is a novel phosphoinositide-binding protein modulating tight junctions and favoring cell migration.
TRAF4 is a novel phosphoinositide-binding protein modulating tight junctions and favoring cell migration.
PLoS biology (2013-12-07)
Adrien Rousseau, Alastair G McEwen, Pierre Poussin-Courmontagne, Didier Rognan, Yves Nominé, Marie-Christine Rio, Catherine Tomasetto, Fabien Alpy
ZUSAMMENFASSUNG
Tumor necrosis factor (TNF) receptor-associated factor 4 (TRAF4) is frequently overexpressed in carcinomas, suggesting a specific role in cancer. Although TRAF4 protein is predominantly found at tight junctions (TJs) in normal mammary epithelial cells (MECs), it accumulates in the cytoplasm of malignant MECs. How TRAF4 is recruited and functions at TJs is unclear. Here we show that TRAF4 possesses a novel phosphoinositide (PIP)-binding domain crucial for its recruitment to TJs. Of interest, this property is shared by the other members of the TRAF protein family. Indeed, the TRAF domain of all TRAF proteins (TRAF1 to TRAF6) is a bona fide PIP-binding domain. Molecular and structural analyses revealed that the TRAF domain of TRAF4 exists as a trimer that binds up to three lipids using basic residues exposed at its surface. Cellular studies indicated that TRAF4 acts as a negative regulator of TJ and increases cell migration. These functions are dependent from its ability to interact with PIPs. Our results suggest that TRAF4 overexpression might contribute to breast cancer progression by destabilizing TJs and favoring cell migration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
16:0–18:1 PE, Avanti Research - A Croda Brand
Avanti
18:1 PA, Avanti Research - A Croda Brand
Avanti
18:1 PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand
Avanti
18:1 PA, Avanti Research - A Croda Brand
Avanti
18:1 PI(3,4,5)P3, 1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder
Avanti
18:1 PI(3,5)P2, Avanti Research - A Croda Brand