Merck
  • Human tenocytes are stimulated to proliferate by acetylcholine through an EGFR signalling pathway.
Human tenocytes are stimulated to proliferate by acetylcholine through an EGFR signalling pathway.
Cell and tissue research (2012-12-06)
Gloria Fong, Ludvig J Backman, Gustav Andersson, Alexander Scott, Patrik Danielson
ZUSAMMENFASSUNG
Studies of human patellar and Achilles tendons have shown that primary tendon fibroblasts (tenocytes) not only have the capacity to produce acetylcholine (ACh) but also express muscarinic ACh receptors (mAChRs) through which ACh can exert its effects. In patients with tendinopathy (chronic tendon pain) with tendinosis, the tendon tissue is characterised by hypercellularity and angiogenesis, both of which might be influenced by ACh. In this study, we have tested the hypothesis that ACh increases the proliferation rate of tenocytes through mAChR stimulation and have examined whether this mechanism operates via the extracellular activation of the epidermal growth factor receptor (EGFR), as shown in other fibroblastic cells. By use of primary human tendon cell cultures, we identified cells expressing vimentin, tenomodulin and scleraxis and found that these cells also contained enzymes related to ACh synthesis and release (choline acetyltransferase and vesicular acetylcholine transporter). The cells furthermore expressed mAChRs of several subtypes. Exogenously administered ACh stimulated proliferation and increased the viability of tenocytes in vitro. When the cells were exposed to atropine (an mAChR antagonist) or the EGFR inhibitor AG1478, the proliferative effect of ACh decreased. Western blot revealed increased phosphorylation, after ACh stimulation, for both EGFR and the extracellular-signal-regulated kinases 1 and 2. Given that tenocytes have been shown to produce ACh and express mAChRs, this study provides evidence of a possible autocrine loop that might contribute to the hypercellularity seen in tendinosis tendon tissue.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Cholin-Acetyltransferase-Antikörper, Chemicon®, from goat
Sigma-Aldrich
Proteasehemmer-Cocktail, for use in tissue culture media, DMSO solution
Sigma-Aldrich
GM 6001, InSolution, ≥95%, broad-spectrum inhibitor of MMPs
Sigma-Aldrich
InSolution AG 1478, CAS-Nr. 175178-82-2, Calbiochem, InSolution, ≥98%, inhibitor EGF receptor tyrosine kinase