Merck
  • Unconventionally secreted effectors of two filamentous pathogens target plant salicylate biosynthesis.
Unconventionally secreted effectors of two filamentous pathogens target plant salicylate biosynthesis.
Nature communications (2014-08-27)
Tingli Liu, Tianqiao Song, Xiong Zhang, Hongbo Yuan, Liming Su, Wanlin Li, Jing Xu, Shiheng Liu, Linlin Chen, Tianzi Chen, Meixiang Zhang, Lichuan Gu, Baolong Zhang, Daolong Dou
ZUSAMMENFASSUNG
Plant diseases caused by fungi and oomycetes pose an increasing threat to food security and ecosystem health worldwide. These filamentous pathogens, while taxonomically distinct, modulate host defense responses by secreting effectors, which are typically identified based on the presence of signal peptides. Here we show that Phytophthora sojae and Verticillium dahliae secrete isochorismatases (PsIsc1 and VdIsc1, respectively) that are required for full pathogenesis. PsIsc1 and VdIsc1 can suppress salicylate-mediated innate immunity in planta and hydrolyse isochorismate in vitro. A conserved triad of catalytic residues is essential for both functions. Thus, the two proteins are isochorismatase effectors that disrupt the plant salicylate metabolism pathway by suppressing its precursor. Furthermore, these proteins lack signal peptides, but exhibit characteristics that lead to unconventional secretion. Therefore, this secretion pathway is a novel mechanism for delivering effectors and might play an important role in host-pathogen interactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-HA-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper der Maus, clone AC-40, purified from hybridoma cell culture