Merck
Transcription factors FOXG1 and Groucho/TLE promote glioblastoma growth.
Nature communications (2013-12-21)
Federica Verginelli, Alessandro Perin, Rola Dali, Karen H Fung, Rita Lo, Pierluigi Longatti, Marie-Christine Guiot, Rolando F Del Maestro, Sabrina Rossi, Umberto di Porzio, Owen Stechishin, Samuel Weiss, Stefano Stifani
ZUSAMMENFASSUNG
Glioblastoma (GBM) is the most common and deadly malignant brain cancer, with a median survival of <2 years. GBM displays a cellular complexity that includes brain tumour-initiating cells (BTICs), which are considered as potential key targets for GBM therapies. Here we show that the transcription factors FOXG1 and Groucho/TLE are expressed in poorly differentiated astroglial cells in human GBM specimens and in primary cultures of GBM-derived BTICs, where they form a complex. FOXG1 knockdown in BTICs causes downregulation of neural stem/progenitor and proliferation markers, increased replicative senescence, upregulation of astroglial differentiation genes and decreased BTIC-initiated tumour growth after intracranial transplantation into host mice. These effects are phenocopied by Groucho/TLE knockdown or dominant inhibition of the FOXG1:Groucho/TLE complex. These results provide evidence that transcriptional programmes regulated by FOXG1 and Groucho/TLE are important for BTIC-initiated brain tumour growth, implicating FOXG1 and Groucho/TLE in GBM tumourigenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoklonaler Anti-GFAP-Antikörper (Glial Fibrillary Acidic Protein, Saures Gliafaserprotein) in Maus hergestellte Antikörper, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Olig2-Antikörper, Klon 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Monoklonaler Anti-S-100-Antikörper (β-Untereinheit) in Maus hergestellte Antikörper, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-Nestin-Antikörper, Klon 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutaminsynthetase-Antikörper, Klon GS-6, clone GS-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Galactocerebrosid-Antikörper, Klon mGalC, clone mGalC, Chemicon®, from mouse
Sigma-Aldrich
Anti-BMI-1-Antikörper, Klon F6, clone F6, Upstate®, from mouse
Sigma-Aldrich
Anti-CNPase-Antikörper, Klon 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Foxg1