Merck
  • Elucidation of the interaction mechanism with liposomes of gH625-peptide functionalized dendrimers.
Elucidation of the interaction mechanism with liposomes of gH625-peptide functionalized dendrimers.
PloS one (2014-11-26)
Annarita Falanga, Rossella Tarallo, Thomas Carberry, Massimiliano Galdiero, Marcus Weck, Stefania Galdiero
ZUSAMMENFASSUNG
We have demonstrated that amide-based dendrimers functionalized with the membrane-interacting peptide gH625 derived from the herpes simplex virus type 1 (HSV-1) envelope glycoprotein H enter cells mainly through a non-active translocation mechanism. Herein, we investigate the interaction between the peptide-functionalized dendrimer and liposomes composed of PC/Chol using fluorescence spectroscopy, isothermal titration calorimetry, and surface plasmon resonance to get insights into the mechanism of internalization. The affinity for the membrane bilayer is very high and the interaction between the peptide-dendrimer and liposomes took place without evidence of pore formation. These results suggest that the presented peptidodendrimeric scaffold may be a promising material for efficient drug delivery.