Merck
  • Crif1 Promotes Adipogenic Differentiation of Bone Marrow Mesenchymal Stem Cells After Irradiation by Modulating the PKA/CREB Signaling Pathway.
Crif1 Promotes Adipogenic Differentiation of Bone Marrow Mesenchymal Stem Cells After Irradiation by Modulating the PKA/CREB Signaling Pathway.
Stem cells (Dayton, Ohio) (2015-04-08)
Xi Zhang, Lixin Xiang, Qian Ran, Yao Liu, Yang Xiang, Yanni Xiao, Li Chen, Fengjie Li, Jiang F Zhong, Zhongjun Li
ZUSAMMENFASSUNG
Dysfunction of the hematopoietic microenvironment is the main obstacle encountered during hematopoiesis reconstruction in patients with acute hematopoietic radiation syndrome. Bone marrow mesenchymal stem cells (BM-MSCs) play a crucial supporting role in hematopoiesis by maintaining the balance between adipogenic and osteogenic differentiation. In this study, we found that irradiation decreased the colony-forming efficiency of BM-MSCs and impaired the balance between adipogenic and osteogenic differentiation. Following irradiation, BM-MCSs became strongly predisposed to adipogenesis, as evidenced by increased oil red O staining and elevated mRNA and protein levels of the adipogenic markers and transcription factors PPARγ and AP2. Overexpression of the essential adipogenesis regulator Crif1 in BM-MSCs promoted adipogenesis after irradiation exposure by upregulating adipogenesis-related genes, including C/EBPβ, PPARγ, and AP2. We found that Crif1 promoted the phosphorylation of cAMP response element binding protein (CREB) through direct interaction with protein kinase A (PKA)-α. Phosphorylation of CREB was inhibited in Crif1-knockdown BM-MSCs even in the presence of a PKA agonist (db-cAMP) and could be suppressed in Crif1-overexpressing BM-MSCs by a PKAα inhibitor (H-89). These results suggest that Crif1 is an indispensable regulator of PKAα cat that modulates the PKA/CREB signaling pathway to promote adipogenic differentiation of BM-MSCs after irradiation.